4-Ethyl-5-fluoro-6-hydroxypyrimidine ≥98%

Supplier: Matrix Scientific

023810-5G 023810-25G
101816-804EA 49.23 USD
101816-804 101816-806
4-Ethyl-5-fluoro-6-hydroxypyrimidine ≥98%
4-Ethyl-5-fluoro-6-hydroxypyrimidine

Hazard: Irritant

Formula: C₆H₇FN₂O
MW: 142.13 g/mol
MDL Number: MFCD08063343
CAS Number: 137234-87-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR