5-Bromo-2-chloroisonicotinic acid ≥98%

Supplier: Matrix Scientific

023772-1G 023772-5G 023772-25G
101816-696EA 23.08 USD
101816-696 101816-698 101816-700
5-Bromo-2-chloroisonicotinic acid ≥98%
5-Bromo-2-chloroisonicotinic acid

Hazard: Irritant/Light Sensitive/Stored under Argon

Formula: C₆H₃BrClNO₂
MW: 236.4500 g/mol
Melting Pt: 220…224 °C
MDL Number: MFCD07375114
CAS Number: 886365-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR