2-Cyano-3-methoxypyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Methoxypicolinonitrile, 3-methoxypyridine-2-carbonitrile
023712-5G
023712-25G
101816-544EA
190.77
USD
101816-544
101816-546
2-Cyano-3-methoxypyridine ≥98%
2-Cyano-3-methoxypyridine
Hazard: Irritant
Formula:
C₇H₆N₂O MW: 134.14 g/mol |
MDL Number:
MFCD07374875 CAS Number: 24059-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...