4-(4-Fluorophenyl)piperidine hydrochloride ≥97%
Supplier: Matrix Scientific
021755-1G
021755-5G
101812-678EA
60
USD
101812-678
101812-680
4-(4-Fluorophenyl)piperidine hydrochloride ≥97%
4-(4-Fluorophenyl)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅ClFN MW: 215.7000 g/mol |
MDL Number:
MFCD01632528 CAS Number: 6716-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...