Morpholine-4-sulfonyl chloride ≥95%
Supplier: Matrix Scientific
021511-1G
021511-5G
101812-220EA
21.54
USD
101812-220
102551-028
Morpholine-4-sulfonyl chloride ≥95%
Morpholine-4-sulfonyl chloride
Hazard: Irritant
Formula:
C₄H₈ClNO₃S |
MDL Number:
MFCD03249981 CAS Number: 1828-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...