4-Fluoro-3-methylbenzenesulfonyl chloride ≥95%

Supplier: Matrix Scientific

021483-1G 021483-5G
101812-166EA 1615.38 USD
101812-166 101812-168
4-Fluoro-3-methylbenzenesulfonyl chloride ≥95%
4-Fluoro-3-methylbenzenesulphonyl chloride

Hazard: Irritant

Formula: C₇H₆ClFO₂S
MW: 208.63 g/mol
MDL Number: MFCD01365818
CAS Number: 629672-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR