2,3-Dibromo-5-fluoropyridine ≥98%

Supplier: Matrix Scientific

021432-1G 021432-5G
101812-090EA 23.08 USD
101812-090 101812-092
2,3-Dibromo-5-fluoropyridine ≥98%
2,3-Dibromo-5-fluoropyridine

Hazard: Irritant/Light Sensitive/Refrigerate/Stored under Argon

Formula: C₅H₂Br₂FN
MDL Number: MFCD08532501
CAS Number: 878207-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR