4-Bromo-3,5-dichloropyridine ≥98%

Supplier: Matrix Scientific

021431-5G 021431-25G
101812-086EA 29.23 USD
101812-086 101812-088
4-Bromo-3,5-dichloropyridine ≥98%
4-Bromo-3,5-dichloropyridine

Hazard: Irritant

Formula: C₅H₂BrCl₂N
MDL Number: MFCD06797978
CAS Number: 343781-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR