2,3,5-Tribromopyridine ≥98%

Supplier: Matrix Scientific

021429-5G 021429-25G
101812-078EA 56.92 USD
101812-078 101812-080
2,3,5-Tribromopyridine ≥98%
2,3,5-Tribromopyridine

Hazard: Irritant/Moisture Sensitive

Formula: C₅H₂Br₃N
MW: 315.7881 g/mol
MDL Number: MFCD00233999
CAS Number: 75806-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR