4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol ≥98%

Supplier: Matrix Scientific

Synonyms: (2,3,5,6-Tetrafluoro-4-(methoxymethyl)phenyl)methanol, 4-(Methoxymethyl)-2,3,5,6-tetrafluorobenzyl alcohol

021427-5G 021427-25G 021427-100G
101812-068EA 15.38 USD
101812-068 101812-070 101812-072
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol ≥98%
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol

Hazard: Irritant

Formula: C₉H₈F₄O₂
MW: 224.16 g/mol
MDL Number: MFCD08688060
CAS Number: 83282-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR