5,6-Dibromopyridine-3-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 5,6-Dibromonicotinic acid

021417-1G 021417-5G
101812-042EA 60 USD
101812-042 101812-044
5,6-Dibromopyridine-3-carboxylic acid ≥98%
5,6-Dibromopyridine-3-carboxylic acid

Hazard: Irritant

Formula: C₆H₃Br₂NO₂
MW: 280.91 g/mol
MDL Number: MFCD01927099
CAS Number: 29241-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR