2-Bromo-5-(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

021399-5G 021399-25G
101811-974EA 73.85 USD
101811-974 101811-976
2-Bromo-5-(trifluoromethyl)benzoic acid ≥98%
2-Bromo-5-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₈H₄BrF₃O₂
MW: 269.0154 g/mol
MDL Number: MFCD06208253
CAS Number: 1483-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR