2,3-Dibromo-5-chloropyridine ≥98%

Supplier: Matrix Scientific

021391-5G 021391-25G
101811-942EA 44.62 USD
101811-942 101811-944
2,3-Dibromo-5-chloropyridine ≥98%
2,3-Dibromo-5-chloropyridine

Hazard: Irritant/Store in Freezer

Formula: C₅H₂Br₂ClN
MW: 271.3400 g/mol
Melting Pt: 39…43 °C
MDL Number: MFCD08687254
CAS Number: 137628-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR