5-Fluoro-2-methoxyphenylboronic acid ≥98%

Supplier: Matrix Scientific

021383-5G 021383-25G
101811-918EA 21.54 USD
101811-918 101811-920
5-Fluoro-2-methoxyphenylboronic acid ≥98%
5-Fluoro-2-methoxyphenylboronic acid

Hazard: Irritant

Formula: C₇H₈BFO₃
MDL Number: MFCD01863526
CAS Number: 179897-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR