5-Chloro-3-cyanopyridine ≥96%

Supplier: Matrix Scientific

Synonyms: 5-Chloronicotinonitrile

021382-1G 021382-5G
101811-914EA 58.46 USD
101811-914 101811-916
5-Chloro-3-cyanopyridine ≥96%
5-Chloro-3-cyanopyridine

Hazard: Irritant

Formula: C₆H₃ClN₂
MW: 138.56 g/mol
MDL Number: MFCD03411334
CAS Number: 51269-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR