3-Hydroxypyridine-2-carbonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Cyano-3-hydroxypyridine, 3-Hydroxypicolinonitrile
021371-5G
021371-25G
101811-878EA
120
USD
101811-878
101811-880
3-Hydroxypyridine-2-carbonitrile ≥98%
3-Hydroxypyridine-2-carbonitrile
Hazard: Irritant
Formula:
C₆H₄N₂O MW: 120.1088 g/mol |
MDL Number:
MFCD01646135 CAS Number: 932-35-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...