5-Fluoro-2-methoxybenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Fluoro-2-methoxybenzene-1-sulphonyl chloride

021294-1G 021294-5G
101811-716EA 33.85 USD
101811-716 101811-718
5-Fluoro-2-methoxybenzenesulfonyl chloride ≥97%
5-Fluoro-2-methoxybenzenesulfonyl chloride

Hazard: Corrosive

Formula: C₇H₆ClFO₃S
MDL Number: MFCD03618459
CAS Number: 67475-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR