3-Amino-6-bromo-1H-indazole

Supplier: Matrix Scientific

Synonyms: 6-Bromo-1H-indazol-3-amine

021271-1G 021271-5G
101811-674EA 36.92 USD
101811-674 101811-676
3-Amino-6-bromo-1H-indazole
3-Amino-6-bromo-1H-indazole

Hazard: Irritant

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Melting Pt: 237-240°C
Density: 0.856
MDL Number: MFCD05665872
CAS Number: 404827-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR