2-Amino-4'-chloroacetophenone hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Chlorophenacylamine hydrochloride
021268-1G
021268-5G
101811-666EA
36.92
USD
101811-666
101811-668
2-Amino-4'-chloroacetophenone hydrochloride ≥97%
2-Amino-4'-chloroacetophenone hydrochloride
Hazard: Irritant
Formula:
C₈H₉Cl₂NO |
MDL Number:
MFCD00193061 CAS Number: 5467-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...