3-Ethyl-5-methylisoxazole-4-carboxylic acid

Supplier: Matrix Scientific

021203-1G 021203-5G
101811-544EA 61.54 USD
101811-544 101811-546
3-Ethyl-5-methylisoxazole-4-carboxylic acid
3-Ethyl-5-methylisoxazole-4-carboxylic acid

Hazard: Irritant

Formula: C₇H₉NO₃
MW: 155.15 g/mol
MDL Number: MFCD03422293
CAS Number: 17147-85-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR