4-(Bromomethyl)-2,3,5,6-tetrafluorobenzotrifluoride

Supplier: Matrix Scientific

Synonyms: 2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide

021047-1G 021047-5G
101811-264EA 40 USD
101811-264 101811-266
4-(Bromomethyl)-2,3,5,6-tetrafluorobenzotrifluoride
4-(Bromomethyl)-2,3,5,6-tetrafluorobenzotrifluoride

Hazard: Irritant/Lachrymator

Formula: C₈H₂BrF₇
MW: 311 g/mol
MDL Number: MFCD00191855
CAS Number: 76437-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR