2-Amino-3,5-dichloro-6-methylpyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Amino-3,5-dichloro-2-methylpyridine

021045-5G 021045-25G
101811-256EA 36.92 USD
101811-256 101811-258
2-Amino-3,5-dichloro-6-methylpyridine ≥98%
2-Amino-3,5-dichloro-6-methylpyridine

Hazard: Irritant

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
MDL Number: MFCD00129029
CAS Number: 22137-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR