3-Cyclohexene-1-carboxylic acid ≥97%

Supplier: Matrix Scientific

011467-5G 011467-25G 011467-100G
101800-704EA 15.38 USD
101800-704 101800-706 101800-708
3-Cyclohexene-1-carboxylic acid ≥97%
3-Cyclohexene-1-carboxylic acid

TCA listed: T
Hazard: Irritant

Formula: C₇H₁₀O₂
Boiling Pt: 234°C
Melting Pt: 17 °C
Density: 1.0981/15°C
MDL Number: MFCD00013781
CAS Number: 4771-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR