Ethyl-2-amino-4-phenylthiophene-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-Amino-4-phenylthiophene-3-carboxylate

011398-1G 011398-5G
101800-570EA 32.31 USD
101800-570 101800-572
Ethyl-2-amino-4-phenylthiophene-3-carboxylate ≥95%
Ethyl-2-amino-4-phenylthiophene-3-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₃NO₂S
Melting Pt: 98 °C
MDL Number: MFCD00126391
CAS Number: 4815-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR