4-(Chloromethyl)thiazole hydrochloride

Supplier: Matrix Scientific

011306-1G 011306-5G
101800-394EA 20 USD
101800-394 101800-396
4-(Chloromethyl)thiazole hydrochloride
4-(Chloromethyl)thiazole hydrochloride

Hazard: Irritant

Formula: C₄H₄ClNS
MW: 170.05 g/mol
Melting Pt: 166…167 °C (subl.)
MDL Number: MFCD02094738
CAS Number: 7709-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR