2-Methoxy-5-pyridinecarboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Methoxynicotinic acid

011264-5G 011264-25G
101800-270EA 30.77 USD
101800-270 101800-272
2-Methoxy-5-pyridinecarboxylic acid ≥98%
2-Methoxy-5-pyridinecarboxylic acid

Hazard: Irritant

Formula: C₇H₇NO₃
Melting Pt: 171…172 °C
MDL Number: MFCD04115721
CAS Number: 66572-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR