3,4-Dichloro-6-(trifluoromethyl)toluene ≥98%

Supplier: Matrix Scientific

Synonyms: 4,5-Dichloro-2-methylbenzotrifluoride

011258-5G 011258-25G
101800-248EA 63.08 USD
101800-248 101800-250
3,4-Dichloro-6-(trifluoromethyl)toluene ≥98%
3,4-Dichloro-6-(trifluoromethyl)toluene

Hazard: Irritant

Formula: C₈H₅Cl₂F₃
MW: 229.03 g/mol
Boiling Pt: 97…100 °C (23 Torr)
MDL Number: MFCD06246858
CAS Number: 74483-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR