3-Chloro-4-methylpyridine ≥98%

Supplier: Matrix Scientific

011255-5G 011255-25G
101800-242EA 100 USD
101800-242 102550-960
3-Chloro-4-methylpyridine ≥98%
3-Chloro-4-methylpyridine

Hazard: Irritant/Irritant-Harmful

Formula: C₆H₆ClN
Boiling Pt: 178 °C
MDL Number: MFCD04114245
CAS Number: 72093-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR