Ethyl-2-chloroisonicotinate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-Chloropyridine-4-carboxylate
011241-5G
011241-25G
101800-198EA
21.54
USD
101800-198
101800-200
Ethyl-2-chloroisonicotinate ≥97%
Ethyl 2-chloroisonicotinate
Hazard: Irritant
Formula:
C₈H₈ClNO₂ MW: 185.6076 g/mol |
MDL Number:
MFCD04114259 CAS Number: 54453-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...