Ethyl-4,4-difluorocyclohexanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 4,4-difluorocyclohexanecarboxylate

011233-1G 011233-5G 011233-25G
101800-178EA 16.92 USD
101800-178 101800-180 101800-182
Ethyl-4,4-difluorocyclohexanecarboxylate ≥97%
Ethyl-4,4-difluorocyclohexanecarboxylate

Hazard: Irritant

Formula: C₉H₁₄F₂O₂
MW: 192.21 g/mol
Boiling Pt: 84-86°C/8mmHg
MDL Number: MFCD03788494
CAS Number: 178312-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR