5-Chloropicolinonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-2-cyanopyridine

011186-5G 011186-25G
101800-114EA 35.38 USD
101800-114 101800-116
5-Chloropicolinonitrile ≥95%
5-Chloropicolinonitrile

Hazard: Irritant

Formula: C6H3ClN2
MW: 138.55 g/mol
Boiling Pt: 110°C/3 Torr
Melting Pt: 83…84 °C
MDL Number: MFCD03788835
CAS Number: 89809-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR