3-Nitrothioanisole ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(methylsulphanyl)-3-nitrobenzene

011170-25G 011170-100G
101800-060EA 369.23 USD
101800-060 101800-062
3-Nitrothioanisole ≥97%
3-Nitrothioanisole

Hazard: Irritant/Stench

Formula: C₇H₇NO₂S
Boiling Pt: 133-134°C/4 Torr
Melting Pt: 14°C
MDL Number: MFCD00024501
CAS Number: 2524-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR