3-Fluorothioanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-3-(methylthio)benzene, 3-Fluorophenyl methyl sulphide

011164-5G 011164-25G 011164-100G
101800-032EA 36.92 USD
101800-032 101800-034 101800-036
3-Fluorothioanisole ≥98%
3-Fluorothioanisole

Hazard: Irritant/Stench

Formula: C₇H₇FS
Boiling Pt: 70-71°C/13 Torr
MDL Number: MFCD03789131
CAS Number: 658-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR