2-Bromo-4,5-difluorobenzoic acid ≥97%

Supplier: Matrix Scientific

011150-5G 011150-25G
101799-996EA 53.85 USD
101799-996 101799-998
2-Bromo-4,5-difluorobenzoic acid ≥97%
2-Bromo-4,5-difluorobenzoic acid

Hazard: Irritant

Formula: C₇H₃BrF₂O₂
MDL Number: MFCD04973422
CAS Number: 64695-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR