6-Bromonicotinonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-5-cyanopyridine

011143-5G 011143-25G
101799-992EA 35.38 USD
101799-992 101799-994
6-Bromonicotinonitrile ≥97%
6-Bromonicotinonitrile

Hazard: Irritant

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Melting Pt: 116…118 °C
MDL Number: MFCD00234141
CAS Number: 139585-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR