4-Amino-α,α,α-trifluoro-m-toluic acid ≥98%
Supplier: Matrix Scientific
011141-5G
011141-25G
101799-988EA
44.62
USD
101799-988
101799-990
4-Amino-α,α,α-trifluoro-m-toluic acid ≥98%
4-Amino-α,α,α-trifluoro-m-toluic acid
Hazard: Irritant
Formula:
C₈H₆F₃NO₂ Melting Pt: 204…206 °C Density: 1.489 |
MDL Number:
MFCD03407960 CAS Number: 400-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...