2-Chloro-6-(trichloromethyl)pyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
Nitrapyrin
011125-5G
011125-25G
101799-938EA
89.23
USD
101799-938
101799-940
2-Chloro-6-(trichloromethyl)pyridine ≥97%
2-Chloro-6-(trichloromethyl)pyridine
TCA listed: T
Hazard: Irritant
Formula:
C₆H₃Cl₄N MW: 230.897 g/mol Boiling Pt: 136-137°C Melting Pt: 63 °C |
MDL Number:
MFCD00072500 CAS Number: 1929-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...