4-Amino-3,5-dichloro-2,6-difluoropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 3,5-Dichloro-2,6-difluoropyridin-4-amine

011085-5G 011085-25G
101799-804EA 47.69 USD
101799-804 101799-806
4-Amino-3,5-dichloro-2,6-difluoropyridine ≥97%
4-Amino-3,5-dichloro-2,6-difluoropyridine

TCA listed: T
Hazard: Irritant

Formula: C₅H₂Cl₂F₂N₂
MW: 198.99 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00011574
CAS Number: 2840-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR