4-(4-Benzylpiperazin-1-yl)phenylamine

Supplier: Matrix Scientific

Synonyms: 4-(4-Benzylpiperazin-1-yl)aniline

011026-500MG 011026-1G
101799-684EA 27.69 USD
101799-684 102663-580
4-(4-Benzylpiperazin-1-yl)phenylamine
4-(4-Benzylpiperazin-1-yl)phenylamine

Hazard: Irritant

Formula: C₁₇H₂₁N₃
Melting Pt: 136 °C
CAS Number: 16154-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR