2-(Perfluoro-7-methyloctyl)ethyl iodide ≥97%

Supplier: Matrix Scientific

005129-5G 005129-25G
101788-300EA 73.85 USD
101788-300 101788-302
2-(Perfluoro-7-methyloctyl)ethyl iodide ≥97%
2-(Perfluoro-7-methyloctyl)ethyl iodide

Hazard: Irritant

Formula: C₁₁H₄F₁₉I
MDL Number: MFCD00153247
CAS Number: 40678-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR