3,4-Difluoro-5-nitrobenzotrifluoride ≥97%
Supplier: Matrix Scientific
005119-250MG
005119-1G
101788-282EA
63.08
USD
101788-282
101788-284
3,4-Difluoro-5-nitrobenzotrifluoride ≥97%
3,4-Difluoro-5-nitrobenzotrifluoride
Hazard: Irritant
Formula:
C₇H₂F₅NO₂ |
CAS Number:
101646-01-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...