(2,4-Dichloro-5-fluorophenyl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-(2,4-Dichloro-5-fluorophenyl)acetic acid

005108-1G 005108-5G
101788-260EA 138.46 USD
101788-260 101788-262
(2,4-Dichloro-5-fluorophenyl)acetic acid
(2,4-Dichloro-5-fluorophenyl)acetic acid

Hazard: Irritant

Formula: (C₆H₂Cl₂F)CH₂COOH
MW: 223.03 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD01631383
CAS Number: 10129-77-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR