2,4-Dichloro-5-fluorobenzyl alcohol

Supplier: Matrix Scientific

005106-1G 005106-5G
101788-256EA 40 USD
101788-256 101788-258
2,4-Dichloro-5-fluorobenzyl alcohol
2,4-Dichloro-5-fluorobenzyl alcohol

Hazard: Irritant

Formula: C₇H₅Cl₂FO
MDL Number: MFCD01631380
CAS Number: 219765-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR