2,3-Difluoro-4-methylbenzamide

Supplier: Matrix Scientific

005072-1G 005072-5G
101788-224EA 38.46 USD
101788-224 101788-226
2,3-Difluoro-4-methylbenzamide
2,3-Difluoro-4-methylbenzamide

Hazard: Irritant

Formula: C₈H₇F₂NO
Melting Pt: 158…162 °C
MDL Number: MFCD01631633
CAS Number: 261763-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR