2,3-Difluoro-4-methylanisole

Supplier: Matrix Scientific

005070-1G 005070-5G
101788-216EA 38.46 USD
101788-216 101788-218
2,3-Difluoro-4-methylanisole
2,3-Difluoro-4-methylanisole

Hazard: Irritant

Formula: C₈H₈F₂O
MDL Number: MFCD01631321
CAS Number: 261763-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR