2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde ≥98%
Supplier: Matrix Scientific
Synonyms:
2,2-Difluorobenzo[d][1,3]dioxole-4-carbaldehyde
005063-1G
005063-5G
101788-200EA
90.77
USD
101788-200
101788-202
2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde ≥98%
2,2-Difluoro-1,3-benzodioxole-4-carboxaldehyde
Hazard: Irritant/Moisture Sensitive/Stored under Argon/Store in Freezer
Formula:
C₈H₄F₂O₃ MW: 186.11 g/mol Boiling Pt: 62-64°C/0.7mm |
MDL Number:
MFCD00792419 CAS Number: 119895-68-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...