2,6-Difluoro-m-cresol

Supplier: Matrix Scientific

005020-1G 005020-5G
101788-172EA 27.69 USD
101788-172 101788-174
2,6-Difluoro-m-cresol
2,6-Difluoro-m-cresol

Hazard: Irritant

Formula: C₇H₆F₂O
Boiling Pt: 182 °C
MDL Number: MFCD01631340
CAS Number: 261763-46-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR