2,6-Difluoro-3-methylbenzyl alcohol

Supplier: Matrix Scientific

Synonyms: (2,6-Difluoro-3-methylphenyl)methanol, 2,4-Difluoro-3-(hydroxymethyl)toluene

005017-1G 005017-5G
101788-168EA 44.62 USD
101788-168 101788-170
2,6-Difluoro-3-methylbenzyl alcohol
2,6-Difluoro-3-methylbenzyl alcohol

Hazard: Irritant

Formula: C₈H₈F₂O
MW: 158.145 g/mol
MDL Number: MFCD01631335
CAS Number: 261763-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR