2,6-Difluoro-3-methylbenzamide

Supplier: Matrix Scientific

005014-1G 005014-5G
101788-160EA 43.08 USD
101788-160 101788-162
2,6-Difluoro-3-methylbenzamide
2,6-Difluoro-3-methylbenzamide

Hazard: Irritant

Formula: C₈H₇F₂NO
MDL Number: MFCD01631408
CAS Number: 261763-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR