2,6-Difluoro-3-methylanisole

Supplier: Matrix Scientific

005012-1G 005012-5G
101788-152EA 47.69 USD
101788-152 101788-154
2,6-Difluoro-3-methylanisole
2,6-Difluoro-3-methylanisole

Hazard: Irritant

Formula: C₈H₈F₂O
Boiling Pt: 53 °C (1 mmHg)
MDL Number: MFCD01631341
CAS Number: 261763-33-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR